3-Cyano-1H-indole-4-carboxylic acid 95%
Supplier: Apollo Scientific
OR956107-1G
OR956107-5G
APOSOR956107-1GEA
270
EUR
APOSOR956107-1G
APOSOR956107-5G
3-Cyano-1H-indole-4-carboxylic acid 95%
3-Cyano-1H-indole-4-carboxylic acid
Formula:
C₁₀H₆N₂O₂ MW: 186,167 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07776626 CAS Number: 889942-85-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...