3-Nitro phenyl ethyl sulphide 95%
Supplier: Apollo Scientific
OR956268-1G
OR956268-5G
APOSOR956268-1GEA
270
EUR
APOSOR956268-1G
APOSOR956268-5G
3-Nitro phenyl ethyl sulphide 95%
3-Nitro phenyl ethyl sulphide
Formula:
C₈H₉NO₂S MW: 183,228 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08460160 CAS Number: 34126-43-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...