8-Methoxy-3,4-dihydrobenzo[b]oxepin-5(2H)-one 95%
Supplier: Apollo Scientific
OR956601-1G
OR956601-250MG
APOSOR956601-1GEA
538
EUR
APOSOR956601-1G
APOSOR956601-250MG
8-Methoxy-3,4-dihydrobenzo[b]oxepin-5(2H)-one 95%
8-Methoxy-3,4-dihydrobenzo[b]oxepin-5(2H)-one
Formula:
C₁₁H₁₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD19285566 CAS Number: 98232-51-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...