5-Chloro-2,3-dihydrobenzofuran 95%
Supplier: Apollo Scientific
OR956999-1G
OR956999-250MG
APOSOR956999-1GEA
342
EUR
APOSOR956999-1G
APOSOR956999-250MG
5-Chloro-2,3-dihydrobenzofuran 95%
5-Chloro-2,3-dihydrobenzofuran
Formula:
C₈H₇ClO Storage Temperature: Ambient |
MDL Number:
MFCD01566475 CAS Number: 76429-69-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...