2-[(Ethylamino)methyl]phenol 95%
Supplier: Apollo Scientific
OR957466-1G
OR957466-5G
APOSOR957466-1GEA
400
EUR
APOSOR957466-1G
APOSOR957466-5G
2-[(Ethylamino)methyl]phenol 95%
2-[(Ethylamino)methyl]phenol
Formula:
C₉H₁₃NO MW: 151,206 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09929100 CAS Number: 108206-05-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...