3-Methoxy-4-methyl-1H-1,2,4-triazol-5(4h)-one 95%

Supplier: Apollo Scientific

Synonyms: 3-Methoxy-4-methyl-1H-1,2,4-triazol-5(4H)-one

OR958138-5G
APOSOR958138-5GEA 60.9 EUR
APOSOR958138-5G
3-Methoxy-4-methyl-1H-1,2,4-triazol-5(4h)-one 95%
3-Methoxy-4-methyl-1H-1,2,4-triazol-5(4h)-one
Formula: C₄H₇N₃O₂
MW: 129,117 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09744038
CAS Number: 135302-13-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR