4-(Pyrrolidin-1-yl)pyridine-2-carbonitrile 98%

Supplier: Apollo Scientific

OR959405-1G OR959405-5G
APOSOR959405-1GEA 119 EUR
APOSOR959405-1G APOSOR959405-5G
4-(Pyrrolidin-1-yl)pyridine-2-carbonitrile 98%
4-(Pyrrolidin-1-yl)pyridine-2-carbonitrile
Formula: C₁₀H₁₁N₃
Storage Temperature: Ambient
MDL Number: MFCD08062860
CAS Number: 127680-86-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR