N-Ethyl 4-chloropicolinamide 98%

Supplier: Apollo Scientific

OR959441-10G OR959441-1G OR959441-5G
APOSOR959441-10GEA 754 EUR
APOSOR959441-10G APOSOR959441-1G APOSOR959441-5G
N-Ethyl 4-chloropicolinamide 98%
N-Ethyl 4-chloropicolinamide
Formula: C₈H₉ClN₂O
MW: 184,623 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08705738
CAS Number: 604813-07-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR