N-Propyl 4-chloropicolinamide 98%
Supplier: Apollo Scientific
OR959443-1G
OR959443-5G
APOSOR959443-1GEA
168
EUR
APOSOR959443-1G
APOSOR959443-5G
N-Propyl 4-chloropicolinamide 98%
N-Propyl 4-chloropicolinamide
Formula:
C₉H₁₁ClN₂O MW: 198,649 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11177055 CAS Number: 694498-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...