N-tert-Butyl-2-chloropyrimidin-4-amine 98%
Supplier: Apollo Scientific
OR959493-1G
OR959493-5G
APOSOR959493-1GEA
219
EUR
APOSOR959493-1G
APOSOR959493-5G
N-tert-Butyl-2-chloropyrimidin-4-amine 98%
N-tert-Butyl-2-chloropyrimidin-4-amine
Formula:
C₈H₁₂ClN₃ MW: 185,654 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09753659 CAS Number: 876521-19-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...