2,5-Dimethoxypyrazine 98%

Supplier: Apollo Scientific

OR959596-1G OR959596-250MG
APOSOR959596-1GEA 841 EUR
APOSOR959596-1G APOSOR959596-250MG
2,5-Dimethoxypyrazine 98%
2,5-Dimethoxypyrazine
Formula: C₆H₈N₂O₂
MW: 140.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18411892
CAS Number: 117856-61-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR