4-Methoxy-3-methylpyridine 98%

Supplier: Apollo Scientific

OR959599-1G OR959599-250MG
APOSOR959599-1GEA 633 EUR
APOSOR959599-1G APOSOR959599-250MG
4-Methoxy-3-methylpyridine 98%
4-Methoxy-3-methylpyridine
Formula: C₇H₉NO
MW: 123,152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06253896
CAS Number: 96609-78-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR