3,5-Dimethoxypyridazine 97%
Supplier: Apollo Scientific
OR959818-1G
OR959818-250MG
APOSOR959818-1GEA
754
EUR
APOSOR959818-1G
APOSOR959818-250MG
3,5-Dimethoxypyridazine 97%
3,5-Dimethoxypyridazine
Formula:
C₆H₈N₂O₂ MW: 140,14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22381712 CAS Number: 2096-20-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...