2-(Butylamino)benzonitrile 98%

Supplier: Apollo Scientific

OR959903-10G OR959903-1G OR959903-5G
APOSOR959903-10GEA 538 EUR
APOSOR959903-10G APOSOR959903-1G APOSOR959903-5G
2-(Butylamino)benzonitrile 98%
2-(Butylamino)benzonitrile
Formula: C₁₁H₁₄N₂
MW: 174,242 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11144682
CAS Number: 5589-61-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR