2-(Cyclopentylamino)benzonitrile 95%

Supplier: Apollo Scientific

OR959904-1G OR959904-5G
APOSOR959904-1GEA 168 EUR
APOSOR959904-1G APOSOR959904-5G
2-(Cyclopentylamino)benzonitrile 95%
2-(Cyclopentylamino)benzonitrile
Formula: C₁₂H₁₄N₂
MW: 186,253 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11144623
CAS Number: 173316-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR