1-Cyclopentylpyrazole 98%

Supplier: Apollo Scientific

OR959961-1G OR959961-5G
APOSOR959961-1GEA 168 EUR
APOSOR959961-1G APOSOR959961-5G
1-Cyclopentylpyrazole 98%
1-Cyclopentylpyrazole
Formula: C₈H₁₂N₂
MW: 136.194 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09701720
CAS Number: 62838-59-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR