2,7-Dimethyl-2,4-dihydro-1,4-benzoxazin-3-one 98%
Supplier: Apollo Scientific
OR960210-1G
OR960210-5G
APOSOR960210-1GEA
290
EUR
APOSOR960210-1G
APOSOR960210-5G
2,7-Dimethyl-2,4-dihydro-1,4-benzoxazin-3-one 98%
2,7-Dimethyl-2,4-dihydro-1,4-benzoxazin-3-one
Formula:
C₁₀H₁₁NO₂ MW: 177.2 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD15730334 CAS Number: 114603-36-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...