3-(3-Methyl-1,2,4-oxadiazol-5-yl)benzonitrile 98%
Supplier: Apollo Scientific
OR960286-1G
OR960286-5G
APOSOR960286-1GEA
321
EUR
APOSOR960286-1G
APOSOR960286-5G
3-(3-Methyl-1,2,4-oxadiazol-5-yl)benzonitrile 98%
3-(3-Methyl-1,2,4-oxadiazol-5-yl)benzonitrile
Formula:
C₁₀H₇N₃O MW: 185,182 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21091814 CAS Number: 1283108-89-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...