3-(4-Chlorophenyl)-4H-1,2,4-triazole 98%

Supplier: Apollo Scientific

OR960296-1G OR960296-5G
APOSOR960296-1GEA 132 EUR
APOSOR960296-1G APOSOR960296-5G
3-(4-Chlorophenyl)-4H-1,2,4-triazole 98%
3-(4-Chlorophenyl)-4H-1,2,4-triazole
Formula: C₈H₆ClN₃
Storage Temperature: Ambient
MDL Number: MFCD19981568
CAS Number: 23195-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR