Ethyl 3-(4-oxopiperidin-1-yl)propanoate 98%

Supplier: Apollo Scientific

OR960344-1G OR960344-250MG
APOSOR960344-1GEA 464 EUR
APOSOR960344-1G APOSOR960344-250MG
Ethyl 3-(4-oxopiperidin-1-yl)propanoate 98%
Ethyl 3-(4-oxopiperidin-1-yl)propanoate
Formula: C₁₀H₁₇NO₃
MW: 199,247 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14631395
CAS Number: 174774-90-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR