(4-Chloro-phenyl)-oxo-acetonitrile 96%
Supplier: Apollo Scientific
OR961763-1G
OR961763-25G
OR961763-5G
APOSOR961763-1GEA
109
EUR
APOSOR961763-1G
APOSOR961763-25G
APOSOR961763-5G
(4-Chloro-phenyl)-oxo-acetonitrile 96%
(4-Chloro-phenyl)-oxo-acetonitrile
Formula:
C₈H₄ClNO MW: 165,576 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06658396 CAS Number: 13014-48-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...