(2S,3R)-3-Methylazetidine-2-carboxylic acid

Supplier: Apollo Scientific

OR961869-1G OR961869-5G
APOSOR961869-1GEA 701 EUR
APOSOR961869-1G APOSOR961869-5G
(2S,3R)-3-Methylazetidine-2-carboxylic acid
(2S,3R)-3-Methylazetidine-2-carboxylic acid
Formula: C₅H₉NO₂
MW: 115,13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19226412
CAS Number: 1932255-43-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR