2-(1H-Imidazol-2-yl)pyridine 98%

Supplier: Apollo Scientific

OR962208-10G OR962208-1G OR962208-5G
APOSOR962208-10GEA 451 EUR
APOSOR962208-10G APOSOR962208-1G APOSOR962208-5G
2-(1H-Imidazol-2-yl)pyridine 98%
2-(1H-Imidazol-2-yl)pyridine
Formula: C₈H₇N₃
MW: 145,161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05863221
CAS Number: 18653-75-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR