2-(2-Hydroxy-ethoxy)-benzoic acid 95%

Supplier: Apollo Scientific

OR962563-1G OR962563-250MG
APOSOR962563-1GEA 364 EUR
APOSOR962563-1G APOSOR962563-250MG
2-(2-Hydroxy-ethoxy)-benzoic acid 95%
2-(2-Hydroxy-ethoxy)-benzoic acid
Formula: C₉H₁₀O₄
MW: 182,173 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01661527
CAS Number: 55211-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR