3-Pyrrolidin-1-ylbenzaldehyde 97%
Supplier: Apollo Scientific
OR962684-1G
OR962684-250MG
APOSOR962684-1GEA
270
EUR
APOSOR962684-1G
APOSOR962684-250MG
3-Pyrrolidin-1-ylbenzaldehyde 97%
3-Pyrrolidin-1-ylbenzaldehyde
Formula:
C₁₁H₁₃NO MW: 175,227 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07772825 CAS Number: 857283-89-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...