tert-Butyl 3-bromobenzylcarbamate 98%

Supplier: Apollo Scientific
Warning

OR962712-1G OR962712-25G OR962712-5G
APOSOR962712-1GEA 29.3 EUR
APOSOR962712-1G APOSOR962712-25G APOSOR962712-5G
tert-Butyl 3-bromobenzylcarbamate 98%
tert-Butyl 3-bromobenzylcarbamate
Formula: C₁₂H₁₆BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD08703139
CAS Number: 171663-13-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR