4-Isopropoxyphenol 97%
Supplier: Apollo Scientific
OR962722-10G
OR962722-1G
OR962722-5G
APOSOR962722-10GEA
227
EUR
APOSOR962722-10G
APOSOR962722-1G
APOSOR962722-5G
4-Isopropoxyphenol 97%
4-Isopropoxyphenol
Formula:
C₉H₁₂O₂ MW: 152,19 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08703155 CAS Number: 7495-77-4 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...