4-Propoxypiperidine hydrochloride 95%

Supplier: Apollo Scientific

OR962881-1G OR962881-5G
APOSOR962881-1GEA 248 EUR
APOSOR962881-1G APOSOR962881-5G
4-Propoxypiperidine hydrochloride 95%
4-Propoxypiperidine hydrochloride
Formula: C₈H₁₇NO·ClH
Storage Temperature: Ambient
MDL Number: MFCD11870087
CAS Number: 903891-77-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR