5-Methoxy-2,3-dihydro-isoindol-1-one 95%
Supplier: Apollo Scientific
Synonyms:
5-Methoxyisoindolin-1-one
OR962968-1G
OR962968-250MG
APOSOR962968-1GEA
207
EUR
APOSOR962968-1G
APOSOR962968-250MG
5-Methoxy-2,3-dihydro-isoindol-1-one 95%
5-Methoxy-2,3-dihydro-isoindol-1-one
Formula:
C₉H₉NO₂ MW: 163,173 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08437636 CAS Number: 22246-66-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...