Cinnoline-4-carboxylic acid 95%

Supplier: Apollo Scientific

OR962973-1G OR962973-250MG
APOSOR962973-1GEA 806 EUR
APOSOR962973-1G APOSOR962973-250MG
Cinnoline-4-carboxylic acid 95%
Cinnoline-4-carboxylic acid
Formula: C₉H₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD00006813
CAS Number: 21905-86-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR