Chloroacetyl-DL-norvaline 95%

Supplier: Apollo Scientific

OR963075-1G OR963075-25G OR963075-5G
APOSOR963075-1GEA 102 EUR
APOSOR963075-1G APOSOR963075-25G APOSOR963075-5G
Chloroacetyl-DL-norvaline 95%
Chloroacetyl-DL-norvaline
Formula: C₇H₁₂ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD00020439
CAS Number: 6940-47-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR