6-Chloro-2H-chromene-3-carbaldehyde 95%
Supplier: Apollo Scientific
OR963325-1G
OR963325-5G
APOSOR963325-1GEA
183
EUR
APOSOR963325-1G
APOSOR963325-5G
6-Chloro-2H-chromene-3-carbaldehyde 95%
6-Chloro-2H-chromene-3-carbaldehyde
Formula:
C₁₀H₇ClO₂ MW: 194,614 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01662416 CAS Number: 57544-34-0 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...