(R)-1-M-Tolylethanamine 95%
Supplier: Apollo Scientific
Synonyms:
(R)-1-m-Tolylethanamine
OR963403-1G
OR963403-250MG
APOSOR963403-1GEA
165
EUR
APOSOR963403-1G
APOSOR963403-250MG
(R)-1-M-Tolylethanamine 95%
(R)-1-M-Tolylethanamine
Formula:
C₉H₁₃N MW: 135.206 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06761972 CAS Number: 138457-19-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...