1-(2-Bromo-4-hydroxyphenyl)ethanone 98%
Supplier: Apollo Scientific
|
Warning
|
OR963788-1G
OR963788-250MG
OR963788-5G
APOSOR963788-1GEA
121
EUR
APOSOR963788-1G
APOSOR963788-250MG
APOSOR963788-5G
1-(2-Bromo-4-hydroxyphenyl)ethanone 98%
1-(2-Bromo-4-hydroxyphenyl)ethanone
Formula:
C₈H₇BrO₂ MW: 215,044 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03428552 CAS Number: 61791-99-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...