3-(3-Hydroxy-propyl)-phenol 96%

Supplier: Apollo Scientific
Warning

OR964224-10G OR964224-1G OR964224-5G
APOSOR964224-10GEA 793 EUR
APOSOR964224-10G APOSOR964224-1G APOSOR964224-5G
3-(3-Hydroxy-propyl)-phenol 96%
3-(3-Hydroxy-propyl)-phenol
Formula: C₉H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD09028718
CAS Number: 1424-74-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR