3,4-Dichloro-5-hydroxyfuran-2(5H)-one 97%

Supplier: Apollo Scientific

OR964429-1G OR964429-5G
APOSOR964429-1GEA 75.4 EUR
APOSOR964429-1G APOSOR964429-5G
3,4-Dichloro-5-hydroxyfuran-2(5H)-one 97%
3,4-Dichloro-5-hydroxyfuran-2(5H)-one
Formula: C₄H₂Cl₂O₃
MW: 168,963 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272415
CAS Number: 766-40-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR