3-[1,2,3]Thiadiazol-4-yl-pyridine 95%

Supplier: Apollo Scientific

OR964493-1G OR964493-250MG
APOSOR964493-1GEA 343 EUR
APOSOR964493-1G APOSOR964493-250MG
3-[1,2,3]Thiadiazol-4-yl-pyridine 95%
3-[1,2,3]Thiadiazol-4-yl-pyridine
Formula: C₇H₅N₃S
MW: 163,2 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11501091
CAS Number: 18212-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR