Ethyl pyridazine-4-carboxylate 95%
Supplier: Apollo Scientific
OR964677-1G
OR964677-250MG
APOSOR964677-1GEA
364
EUR
APOSOR964677-1G
APOSOR964677-250MG
Ethyl pyridazine-4-carboxylate 95%
Ethyl pyridazine-4-carboxylate
Formula:
C₇H₈N₂O₂ MW: 152,151 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09833000 CAS Number: 39123-39-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...