5-Pyridin-3-ylisoxazole-3-carboxylic acid 96%

Supplier: Apollo Scientific

Synonyms: 5-(3-Pyridyl)isoxazole-3-carboxylic Acid

OR964778-100MG OR964778-1G OR964778-250MG
APOSOR964778-100MGEA 53.8 EUR
APOSOR964778-100MG APOSOR964778-1G APOSOR964778-250MG
5-Pyridin-3-ylisoxazole-3-carboxylic acid 96%
5-Pyridin-3-ylisoxazole-3-carboxylic acid
Formula: C₉H₆N₂O₃
MW: 190.156 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07377114
CAS Number: 893638-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR