N-Benzyl-N-(2-methoxyethyl)amine 98%

Supplier: Apollo Scientific

OR964902-100MG OR964902-1G OR964902-250MG
APOSOR964902-100MGEA 183 EUR
APOSOR964902-100MG APOSOR964902-1G APOSOR964902-250MG
N-Benzyl-N-(2-methoxyethyl)amine 98%
N-Benzyl-N-(2-methoxyethyl)amine
Formula: C₁₀H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD01445287
CAS Number: 51353-26-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR