(R)-2-((Methoxycarbonyl)amino)-3-methylbutanoic acid 95%
Supplier: Apollo Scientific
OR965041-1G
OR965041-5G
APOSOR965041-1GEA
49.3
EUR
APOSOR965041-1G
APOSOR965041-5G
(R)-2-((Methoxycarbonyl)amino)-3-methylbutanoic acid 95%
(R)-2-((Methoxycarbonyl)amino)-3-methylbutanoic acid
Formula:
C₇H₁₃NO₄ MW: 175,182 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD21234526 CAS Number: 171567-86-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...