2-Hydroxyethanesulphonic acid 80% in water 97%

Supplier: Apollo Scientific
Danger

Synonyms: Isethionic acid

OR965087-100G OR965087-500G
APOSOR965087-100GEA 102 EUR
APOSOR965087-100G APOSOR965087-500G
2-Hydroxyethanesulphonic acid 80% in water 97%
2-Hydroxyethanesulphonic acid
Formula: C₂H₆O₄S
MW: 126.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00242599
CAS Number: 107-36-8
UN: 3265
ADR: 8 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR