3-Methyl-2(3H)-benzothiazolone 95%
Supplier: Apollo Scientific
OR965197-1G
OR965197-5G
APOSOR965197-1GEA
168
EUR
APOSOR965197-1G
APOSOR965197-5G
3-Methyl-2(3H)-benzothiazolone 95%
3-Methyl-2(3H)-benzothiazolone
Formula:
C₈H₇NOS Storage Temperature: Ambient |
MDL Number:
MFCD00022914 CAS Number: 2786-62-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...