O-(2-vinyloxyethyl)hydroxylamine 98%
Supplier: Apollo Scientific
|
Warning
|
OR965288-1G
OR965288-5G
APOSOR965288-1GEA
241
EUR
APOSOR965288-1G
APOSOR965288-5G
O-(2-vinyloxyethyl)hydroxylamine 98%
O-(2-vinyloxyethyl)hydroxylamine
Formula:
C₄H₉NO₂ MW: 103,12 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD09951926 CAS Number: 391212-29-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...