4-Methyl-1,3-thiazol-2(3H)-one 98%

Supplier: Apollo Scientific

OR965594-100MG OR965594-1G OR965594-250MG
APOSOR965594-100MGEA 168 EUR
APOSOR965594-100MG APOSOR965594-1G APOSOR965594-250MG
4-Methyl-1,3-thiazol-2(3H)-one 98%
4-Methyl-1,3-thiazol-2(3H)-one
Formula: C₄H₅NOS
MW: 115,154 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00970121
CAS Number: 32497-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR