2-(2-Bromoethoxy)propane 97%

Supplier: Apollo Scientific

OR965620-1G OR965620-250MG
APOSOR965620-1GEA 101 EUR
APOSOR965620-1G APOSOR965620-250MG
2-(2-Bromoethoxy)propane 97%
2-(2-Bromoethoxy)propane
Formula: C₅H₁₁BrO
MW: 167,044 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07700570
CAS Number: 54149-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR