4-(4-Methoxyphenyl)pyridine 95%
Supplier: Apollo Scientific
Synonyms:
4-(4-Methoxyphenyl)pyridine
OR965665-1G
OR965665-250MG
APOSOR965665-1GEA
212
EUR
APOSOR965665-1G
APOSOR965665-250MG
4-(4-Methoxyphenyl)pyridine 95%
4-(4-Methoxyphenyl)pyridine
Formula:
C₁₂H₁₁NO MW: 185,222 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00092331 CAS Number: 5938-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...