4-Cyclopropyl-2-methylbenzoic acid 95%

Supplier: Apollo Scientific

OR965829-1G OR965829-250MG
APOSOR965829-1GEA 154 EUR
APOSOR965829-1G APOSOR965829-250MG
4-Cyclopropyl-2-methylbenzoic acid 95%
4-Cyclopropyl-2-methylbenzoic acid
Formula: C₁₁H₁₂O₂
MW: 176.212 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20645932
CAS Number: 909698-10-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR