2-(1-Methyl-1H-benzoimidazol-2-ylamino)-ethanol 96%

Supplier: Apollo Scientific

Synonyms: 2-((1-Methyl-1H-benzo[d]imidazol-2-yl)amino)ethanol

OR965847-1G OR965847-5G
APOSOR965847-1GEA 292 EUR
APOSOR965847-1G APOSOR965847-5G
2-(1-Methyl-1H-benzoimidazol-2-ylamino)-ethanol 96%
2-(1-Methyl-1H-benzoimidazol-2-ylamino)-ethanol
Formula: C₁₀H₁₃N₃O
MW: 191,23 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01136088
CAS Number: 57262-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR