4-(5-Methyl-1,2,4-oxadiazol-3-yl)aniline 97%

Supplier: Apollo Scientific
Warning

OR9660-1G OR9660-250MG
APOSOR9660-1GEA 328 EUR
APOSOR9660-1G APOSOR9660-250MG
4-(5-Methyl-1,2,4-oxadiazol-3-yl)aniline 97%
4-(5-Methyl-1,2,4-oxadiazol-3-yl)aniline
Formula: C₉H₉N₃O
Melting Pt: 113…114 °C
Storage Temperature: Ambient
MDL Number: MFCD08435856
CAS Number: 10185-68-9
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR